Learn More
Medchemexpress LLC HY-13555 10mg , β-Lapachone CAS:4707-32-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1355510MG
Encompass_Preferred
HY-13555 10mg β-Lapachone CAS: 4707-32-8 β-Lapachone (ARQ-501;NSC-26326) is a naturally occurring O-naphthoquinone, acts as a topoisomerase I inhibitor, and induces apoptosis by inhibiting cell cycle progression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9244