Learn More
Sigma Aldrich Fine Chemicals Biosciences B Medical L290 Lab Refrige1EA
Catalog No. 502909022
Encompass
.nbspPrecision - Upright Mid-Size Laboratory Refrigerator.nbsp1 Bottom Drawer.nbsp5 Wire Shelves.nbspDigital Temperature Display.nbspAlarm System w/ Battery Back-Up.nbspUSB Data Port.nbspInternal LED Light
Catalog No. 50-290-9022 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BM991.8326.621EA
.nbspPrecision - Upright Mid-Size Laboratory Refrigerator.nbsp1 Bottom Drawer.nbsp5 Wire Shelves.nbspDigital Temperature Display.nbspAlarm System w/ Battery Back-Up.nbspUSB Data Port.nbspInternal LED Light