Learn More
Andwin Scientific B-MERCAPTOETHANOL 250ML
SDP
Supplier:  Andwin Scientific EM444203
Encompass_Preferred
The 250mL -Mercaptoethanol, also known as Thioglycol, is a clear, colorless liquid chemical that has a pH of 4.5-6 at 68 F or 20C. It can be used as a standard buffer. This product is combustible and is classified as toxic by inhalation and ingestion.
Catalog No. NC9580853