Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-B-Methoxydiisopinocampheylborane

Catalog No. 31704725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-704-725G 25 g
31-704-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 31-704-725G Supplier Sigma Aldrich Supplier No. 31704725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

B-Methoxydiisopinocampheylborane (Ipc2BOMe) is an organoborane compound, which is prepared from excess +a-pinene, borane dimethylsulfide, and methanol via the formation of an intermediate diisocampheylborane. Ipc2BOMe is used as a versatile reagent for the construction of C-C bonds in asymmetric synthesis.Reactant involved in organic synthesis reactions such as:Double allylboration for synthesis of fragments of tetrafibricinAnticancer cytotoxic monorhizopodin synthesisAnnulation of cyclic allylsilanesAsymmetric synthesis of +a-amino-+a-hydroxy acid taxol side chain analogs

Specifications

CAS 85134-98-1
Molecular Formula C21H37BO
Linear Formula C21H37BO
MDL Number MFCD00074809
Quantity 25 g
Synonym (-)-Diisopinocampheylmethoxyborane
Molecular Weight (g/mol) 316.33 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.