Learn More
Medchemexpress LLC B-Naphthoflavone | 100MG
SDP
Catalog No. 5000226587
Encompass_Preferred
-Naphthoflavone is an exogenous aryl hydrocarbon receptor (AHR) ligand -Naphthoflavone can activate AHR to participate in various biological processes including cell growth differentiation apoptosis and metabolism -Naphthoflavone has antioxidant activity and can exert its antioxidant function by regulating the activity of antioxidant enzymes -Naphthoflavone is also a non-carcinogenic CYP1A inducer and can be used to treat aristolochic acid (AAI) induced renal injury[1][2][3]
Catalog No. 50-002-26587 Supplier Medchemexpress LLC Supplier No. HY114740100MG
-Naphthoflavone is an exogenous aryl hydrocarbon receptor (AHR) ligand -Naphthoflavone can activate AHR to participate in various biological processes including cell growth differentiation apoptosis and metabolism -Naphthoflavone has antioxidant activity and can exert its antioxidant function by regulating the activity of antioxidant enzymes -Naphthoflavone is also a non-carcinogenic CYP1A inducer and can be used to treat aristolochic acid (AAI) induced renal injury[1][2][3]