Learn More
Alkali Scientific B Naphthoxyacetic Acid, 500 G
SDP
Supplier:  Alkali Scientific PHT003CP
Encompass
Naphthoxyacetic acid, also known as 2-NAA, is a widely used plant regulator that mimics the action of natural plant auxins to promote rooting and regulate growth. This 500g quantity of naphthoxyacetic acid is ideal for large-scale agricultural studies, particularly in plant propagation, tissue culture, and horticultural experiments. NAA is commonly applied to stimulate root formation in plant cuttings, enhance fruit set, and control vegetative growth. It is especially useful in research related to crop yield optimization, as it can regulate flowering time and fruit size. The product is also used in studies involving the regulation of plant hormones to control plant development at various stages.
Catalog No. 50-006-61216