Learn More
Enzo Life Sciences beta Nerve growth factor (human), (recombinant) (5µg). CAS: 86923-98-0
SDP
Catalog No. 502009350
Encompass_Preferred
Alternative name: beta NGF. Purity: ≥98% (HPLC). Formulation: Lyophilized. No additives. Source: Produced in E. coli. Dimer of two identical subunits (119 aa) linked together by very strong hydrophobic interactions. Long Term Storage: -20°C.
Catalog No. 50-200-9350 Supplier Enzo Life Sciences Supplier No. ALX521005C005
Alternative name: beta NGF. Purity: ≥98% (HPLC). Formulation: Lyophilized. No additives. Source: Produced in E. coli. Dimer of two identical subunits (119 aa) linked together by very strong hydrophobic interactions. Long Term Storage: -20°C.