Learn More
Enzo Life Sciences beta Nerve growth factor (mouse), (recombinant) (5µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2640005
Encompass_Preferred
Alternative name: beta NGF. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA). Source: Produced in E. coli. Contains 121/242 amino acids. . Long Term Storage: -80°C.
Catalog No. 50-201-4080