Learn More
Sigma Aldrich Fine Chemicals Biosciences β-Nicotinamide adenine dinucleotide sodium salt, 20111-18-6, MFCD00148917, 1 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences N06321G
Encompass
Empirical Formula (Hill Notation): C21H26N7NaO14P2, Molecular Weight: 685.41, ≥95% (HPLC), storage temp: −20°C, Synonym: β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide.
Catalog No. NC0935497