Learn More
Selleck Chemical LLC b-Nicotinamide Mononucleotide S5259-500mg
SDP
Catalog No. 503111923
Encompass_Preferred
Nicotinamide mononucleotide ( -NM NMN) one of the key precursors of NAD and products of the NAMPT reaction is converted to NAD by nicotinamide mononucleotide adenylyltransferase
Catalog No. 50-311-1923 Supplier Selleck Chemical LLC Supplier No. S5259500MG
Nicotinamide mononucleotide ( -NM NMN) one of the key precursors of NAD and products of the NAMPT reaction is converted to NAD by nicotinamide mononucleotide adenylyltransferase