Learn More
Selleck Chemical LLC b-Nicotinamide Mononucleotide S5259-1g
SDP
Catalog No. 503112450
Encompass_Preferred
Nicotinamide mononucleotide ( -NM NMN) one of the key precursors of NAD and products of the NAMPT reaction is converted to NAD by nicotinamide mononucleotide adenylyltransferase
Catalog No. 50-311-2450 Supplier Selleck Chemical LLC Supplier No. S52591G
Nicotinamide mononucleotide ( -NM NMN) one of the key precursors of NAD and products of the NAMPT reaction is converted to NAD by nicotinamide mononucleotide adenylyltransferase