Learn More
Creative Diagnostics Bordetella pertussis Fimbriae 2/3, Fim 2 Subunit Molecular Weight 22,500 Da, Fim 3 Subunit Molecular Weight 22,000 Da, Antigen
Catalog No. NC2178141
Encompass
The Bordetella pertussis Fimbriae 2/3 is an essential virulence factor that plays a major role in the adherence of Bordetella pertussis to the epithelial lining of the host respiratory tract. Comprising two distinct fimbriae serotypes (Fim 2 and Fim 3), these extracellular proteins are crucial for the attachment of the bacteria, thereby contributing to the infection process. This product is invaluable for research and development, particularly in the field of pertussis vaccine development.
  • Critical for adherence to host epithelial cells
  • Key for establishing Bordetella pertussis infection
  • Valuable antigen for vaccine research
  • Fimbriae serotypes: Fim 2 and Fim 3
  • Fim 2 subunit molecular weight: 22,500 Da
  • Fim 3 subunit molecular weight: 22,000 Da
  • Subunits dissociate in SDS
  • Antibodies to native Fim do not react well with SDS-denatured subunits
  • Batch-dependent product
This item is not returnable. View return policy
Catalog No. NC2178141 Supplier Creative Diagnostics Supplier No. DAGB126
The Bordetella pertussis Fimbriae 2/3 is an essential virulence factor that plays a major role in the adherence of Bordetella pertussis to the epithelial lining of the host respiratory tract. Comprising two distinct fimbriae serotypes (Fim 2 and Fim 3), these extracellular proteins are crucial for the attachment of the bacteria, thereby contributing to the infection process. This product is invaluable for research and development, particularly in the field of pertussis vaccine development.
  • Critical for adherence to host epithelial cells
  • Key for establishing Bordetella pertussis infection
  • Valuable antigen for vaccine research
  • Fimbriae serotypes: Fim 2 and Fim 3
  • Fim 2 subunit molecular weight: 22,500 Da
  • Fim 3 subunit molecular weight: 22,000 Da
  • Subunits dissociate in SDS
  • Antibodies to native Fim do not react well with SDS-denatured subunits
  • Batch-dependent product