Learn More
Drg International Inc DRG Bordetella Pertussis IgA ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3449R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgA-class antibodies to Bordetella pertussis and Bordetella pertussis toxin in serum and plasma (EDTA or heparin plasma). This product is for Research Use Only. The DRG Bordetella pertussis / toxin IgA ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with Bordetella pertussis and Bordetella pertussis toxin antigen. Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7557