Learn More
Drg International Inc DRG Bordetella Pertussis IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3451R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgM-class antibodies to Bordetella pertussis and Bordetella pertussis toxin in serum and plasma (EDTA- or heparin plasma). This product is for Research Use Only. The DRG Bordetella pertussis / toxin IgM ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, containing hyper-immune anti-human IgG-class antibody to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors.
Catalog No. 50-194-7559