Learn More
Description
- Type M Ultrapure for use with B-Pure Deionization System
- Resin type: Mixed bed
- High purity
Specifications
Specifications
| Capacity | 760 grains as NaCl, 875 |
| Resin Type | Mixed Bed |
| For Use With (Equipment) | B-Pure™ deionization system and analytical lab testing |
| Purity | High |
| Type | Type M Ultrapure Cartridge |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
