Learn More
Thermo Scientific™ B·R·A·H·M·S PCT™ sensitive KRYPTOR™
GSA_VA
Catalog No. 23180002 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Tests
Catalog No. Quantity
23-180-002 50 Tests
1 options
Catalog No. 23-180-002
Add to Cart
Add to Cart
Homogeneous immunoassay (sandwich principle) using TRACE™ technology.
B·R·A·H·M·S PCT™ sensitive KRYPTOR™ is an immunofluorescent assay used to determine the concentration of PCT (procalcitonin) in human serum and plasma.
B·R·A·H·M·S PCT sensitive KRYPTOR is intended for use in conjunction with other laboratory findings and clinical assessments to aid in the risk assessment of critically ill patients on their first day of ICU admission for progression to severe sepsis and septic shock.
Sepsis is a daily challenge in intensive care units. Today various therapeutic strategies are known to improve survival in patients with sepsis. Early assessment is important for determination of the appropriate treatment.
PCT is the prohormone of the hormone calcitonin; Calcitonin is exclusively produced by C-cells of the thyroid gland in response to hormonal stimuli, whereas PCT can be produced by several cell types and many organs in response to pro-inflammatory stimuli, in particular by bacterial products.
In healthy people, plasma PCT concentrations are found to be below 0.1 μg/l. PCT levels rise rapidly after a bacterial infectious insult with systemic consequences. Early after multiple traumas, major surgery, severe burns or in neonates, PCT levels can be elevated independently of an infectious process, but the return to baseline is usually rapid. Viral infections, bacterial colonization, localized infections, allergic disorders, autoimmune diseases and transplant rejection do not usually induce a significant PCT response (values < 0.5 μg/l). Therefore, by evaluating PCT concentrations, the physician may use the findings to aid in the risk assessment for progression to severe sepsis and septic shock.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Immunofluorescent Assay
Quantity 50 Tests
Disclaimers: Some of these products are not cleared in every country. Please contact your sales representative for more information. Product claims may differ from country to country based on regulations and product approvals. Please contact your country representative for further details.