Learn More
Eurofins Discoverx B-Raf (δ1-415), active, 250 µg. (14-530M)
Supplier:  Eurofins Discoverx 14530M
Encompass
N-terminal GST-tagged recombinant, human B-Raf amino acids 416-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8930