Learn More
Medchemexpress LLC B-Raf IN 1 | 950736-05-7 | MFCD25977055 | 10mg
SDP
Catalog No. NC1556797
Encompass_Preferred
B-Raf IN 1 | Putity: 98.08% | MW: 515.53 | 950736-05-7 | MFCD25977055 | 10mg
Catalog No. NC1556797 Supplier Medchemexpress LLC Supplier No. HY1822710MG
B-Raf IN 1 | Putity: 98.08% | MW: 515.53 | 950736-05-7 | MFCD25977055 | 10mg