Learn More
Selleck Chemical LLC B-Raf IN 1
SDP
Catalog No. 5000002916
Encompass_Preferred
B-Raf IN 1 is an inhibitor of Raf wih IC50 values of 24 nM and 25 nM for B-Raf and C-Raf respectively It is selective over 13 other kinases including PKC IKK and PI3K
Catalog No. 50-000-02916 Supplier Selleck Chemical LLC Supplier No. S65382MG
B-Raf IN 1 is an inhibitor of Raf wih IC50 values of 24 nM and 25 nM for B-Raf and C-Raf respectively It is selective over 13 other kinases including PKC IKK and PI3K