Learn More
Eurofins Discoverx B-Raf (V599E), active, 10 µg. (14-557)
Supplier:  Eurofins Discoverx 14557
Encompass
N-terminal GST-tagged recombinant, human B-Raf amino acids 416-end, containing a V599E mutation (also designated V600E). For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8965