Learn More
MSC TESA Brown & Sharpe Bestest Indicator Contact Point, Carbide, 0.040 in. Ball, 1-7/16 in. Length, 1 ea
Catalog No. 5000510420
Encompass
This carbide contact point is designed for use with dial test indicators, providing a reliable interface for precise measurement applications. It features a ball-style tip for versatile surface contact and is manufactured to exacting standards for consistent performance.
  • Carbide material ensures durability and wear resistance
  • Ball end design for effective measurement contact
  • Thread size of M1.4 x 0.3
  • Compatible with specified test indicator models
  • Ball diameter of 1.00 mm (0.0400 in.)
  • Tip length of 36.51 mm (1-7/16 in.)
Catalog No. 50-005-10420 Supplier MSC Supplier No. 96006069
This carbide contact point is designed for use with dial test indicators, providing a reliable interface for precise measurement applications. It features a ball-style tip for versatile surface contact and is manufactured to exacting standards for consistent performance.
  • Carbide material ensures durability and wear resistance
  • Ball end design for effective measurement contact
  • Thread size of M1.4 x 0.3
  • Compatible with specified test indicator models
  • Ball diameter of 1.00 mm (0.0400 in.)
  • Tip length of 36.51 mm (1-7/16 in.)