Learn More
Kyfora Bio Beta-sitostanol 5 MG
Catalog No. 502559793
Encompass_Preferred
β-Sitostanol is a saturated plant sterol belonging to the same family as campesterol and β-sitosterol. This compound is structurally similar to cholesterol and is known to integrate into lipid bilayers, affecting membrane fluidity and function. β-Sitostanol is widely used in scientific research applications, including the formulation of micelles and liposomes. It serves as a valuable tool in cell culture studies for investigating cellular uptake mechanisms, lipid metabolism, and cholesterol absorption.Applications: Micelle formulation, cell culture studies, cellular uptake monitoring
Catalog No. 50-255-9793 Supplier Kyfora Bio Supplier No. 145245
β-Sitostanol is a saturated plant sterol belonging to the same family as campesterol and β-sitosterol. This compound is structurally similar to cholesterol and is known to integrate into lipid bilayers, affecting membrane fluidity and function. β-Sitostanol is widely used in scientific research applications, including the formulation of micelles and liposomes. It serves as a valuable tool in cell culture studies for investigating cellular uptake mechanisms, lipid metabolism, and cholesterol absorption.Applications: Micelle formulation, cell culture studies, cellular uptake monitoring