Learn More
Bellco Glass, Inc. Bench-Top Digital Glass Door Incubator, 115V
SDP
Catalog No. 501946045
Encompass_Preferred
The Bellco Bench Top Incubator was designed for the incubation of cell cultures and bacteria within laboratories where space is a premium. The instrument panel is mounted on top for easy viewing and manipulation. Glass door models allow the viewing of cultures without having toopen the door limiting possible contaminationproblems and temperature fluctuations. Theconvenient interior accessory outlet allows theuse of Bellco 4 and 10 Position Roller Apparatus, Rockers, Shakersand Magnetic Stirrers. Optional stainless steel shelf createsadditional space for storing cell culture plates and flasks. MADE IN THE USA
Catalog No. 50-194-6045 Supplier Bellco Glass, Inc. Supplier No. 772820115
Add to Cart
Add to Cart
Request a Quote
The Bellco Bench Top Incubator was designed for the incubation of cell cultures and bacteria within laboratories where space is a premium. The instrument panel is mounted on top for easy viewing and manipulation. Glass door models allow the viewing of cultures without having toopen the door limiting possible contaminationproblems and temperature fluctuations. Theconvenient interior accessory outlet allows theuse of Bellco 4 and 10 Position Roller Apparatus, Rockers, Shakersand Magnetic Stirrers. Optional stainless steel shelf createsadditional space for storing cell culture plates and flasks. MADE IN THE USA