Learn More
Bellco Glass, Inc. Bench Top Roller Apparatus,230V-2 P BTBII - Base only
SDP
Catalog No. 501945960
Encompass_Preferred
The Bellco Bench top Modular Roller Apparatus is a variable speed two bottle roller apparatus (expandable to six bottles) used for the purpose of producing animal cells or cell products. This small sized unit is ideal for experimental and pilot-scale cell culture. The unit features a heavy-duty DC motor and solid-state electronics for reliable operation. The unit includes All-Position Drive belts so that every roller rotates, thus preventing slippage of light-weight plastic bottles. Bellco’s unique hex-shaped shafts prevent roller slippage and produce positive drive. MADE IN THE USA
Catalog No. 50-194-5960 Supplier Bellco Glass, Inc. Supplier No. 763200230
The Bellco Bench top Modular Roller Apparatus is a variable speed two bottle roller apparatus (expandable to six bottles) used for the purpose of producing animal cells or cell products. This small sized unit is ideal for experimental and pilot-scale cell culture. The unit features a heavy-duty DC motor and solid-state electronics for reliable operation. The unit includes All-Position Drive belts so that every roller rotates, thus preventing slippage of light-weight plastic bottles. Bellco’s unique hex-shaped shafts prevent roller slippage and produce positive drive. MADE IN THE USA