Learn More
Cusabio Technology LLC B-Thromboglobulin B-TG
Catalog No. 5000050491
Encompass_Preferred
The Human B-Thromboglobulin (B-TG) ELISA Kit is designed to quantitatively measure B-TG levels in serum plasma and tissue homogenates using a sandwich ELISA methodology This kit detects B-TG within a dynamic range of 0 78 ng/mL to 50 ng/mL with a sensitivity of 0 195 ng/mL ensuring precise detection of low-abundance samples The assay requires a short incubation period of 1-5 hours and utilizes a sample volume of 50-100 L per test Measurements are performed at an absorbance wavelength of 450 nm providing reliable and reproducible results for research applications Suitable for clinical and laboratory settings this kit offers a robust tool for studying thrombocyte activation and related disorders
Catalog No. 50-000-50491 Supplier Cusabio Technology LLC Supplier No. CSBE07886H
The Human B-Thromboglobulin (B-TG) ELISA Kit is designed to quantitatively measure B-TG levels in serum plasma and tissue homogenates using a sandwich ELISA methodology This kit detects B-TG within a dynamic range of 0 78 ng/mL to 50 ng/mL with a sensitivity of 0 195 ng/mL ensuring precise detection of low-abundance samples The assay requires a short incubation period of 1-5 hours and utilizes a sample volume of 50-100 L per test Measurements are performed at an absorbance wavelength of 450 nm providing reliable and reproducible results for research applications Suitable for clinical and laboratory settings this kit offers a robust tool for studying thrombocyte activation and related disorders