Learn More
LICOR Tubulin Rabbit Polyclonal Antibody for Normalization (100 L)
Supplier:  LICOR 92642211
Encompass_Preferred
The β-Tubulin primary antibody can be used as an internal loading control (ILC) for normalization. The expression of β-tubulin, or any housekeeping protein (HKP), should be validated to ensure that its expression does not change under experimental conditions. Once validated, β-tubulin primary antibodies can be used for the detection of β-tubulin when performing two-color detection. Detect β-Tubulin Rabbit Polyclonal Antibody with IRDye Goat anti-Rabbit or IRDye Donkey anti-Rabbit secondary antibodies.
Catalog No. NC9872627