Learn More
MSC Guardair Vacuum Kit, 55 Gallon, ATEX Certified, 2 in Inlet
Catalog No. 502839847
Encompass
This pneumatic vacuum kit is ATEX-certified for use in hazardous locations, including those with combustible dust. It features electrically bonded components and static conductive attachments to eliminate static electricity. Operating on compressed air with no electric parts, it generates superior vacuum suction and offers significant cost savings.
  • ATEX certified for hazardous locations
  • Energy efficient, consuming up to 70% less compressed air
  • Customizable performance with three injector nozzle sets
  • Intrinsically safe, eliminating electrical sparks and cords
  • Proprietary multi-venturi engine design for superior performance
  • Versatile for both wet and dry applications with liquid shutoff
  • Compatible with existing vacuum components
  • Includes static conductive vacuum and air supply hoses, plus metal accessories
Catalog No. 50-283-9847 Supplier MSC Supplier No. 92383439
This pneumatic vacuum kit is ATEX-certified for use in hazardous locations, including those with combustible dust. It features electrically bonded components and static conductive attachments to eliminate static electricity. Operating on compressed air with no electric parts, it generates superior vacuum suction and offers significant cost savings.
  • ATEX certified for hazardous locations
  • Energy efficient, consuming up to 70% less compressed air
  • Customizable performance with three injector nozzle sets
  • Intrinsically safe, eliminating electrical sparks and cords
  • Proprietary multi-venturi engine design for superior performance
  • Versatile for both wet and dry applications with liquid shutoff
  • Compatible with existing vacuum components
  • Includes static conductive vacuum and air supply hoses, plus metal accessories