Learn More
MSC Guardair PowerQUAD ATEX Vacuum, 30 gal, 1.5 in, 1 ea
Catalog No. 502870199
Encompass
The Guardair PowerQUAD 30-Gallon ATEX Vacuum Kit is a pneumatic vacuum system engineered for exceptional performance in hazardous locations. It is ATEX-certified and designed for vacuuming combustible dust, chips, debris, liquids, or sludge, ensuring safety by eliminating static electricity at the source.
  • ATEX certified for use in potentially explosive atmospheres
  • Consumes up to 70% less compressed air for energy efficiency
  • Customizable performance with three color-coded injector nozzle sets
  • Intrinsically safe operation with no electric motors or cords
  • Features a rugged, diecast aluminum, 2-stage, multi-venturi engine
  • Wet/dry capable with a liquid shutoff assembly
  • Backwards compatible with existing pneumatic vacuum components
  • Electrically bonded components and static conductive attachments eliminate static electricity
Catalog No. 50-287-0199 Supplier MSC Supplier No. 92383454
The Guardair PowerQUAD 30-Gallon ATEX Vacuum Kit is a pneumatic vacuum system engineered for exceptional performance in hazardous locations. It is ATEX-certified and designed for vacuuming combustible dust, chips, debris, liquids, or sludge, ensuring safety by eliminating static electricity at the source.
  • ATEX certified for use in potentially explosive atmospheres
  • Consumes up to 70% less compressed air for energy efficiency
  • Customizable performance with three color-coded injector nozzle sets
  • Intrinsically safe operation with no electric motors or cords
  • Features a rugged, diecast aluminum, 2-stage, multi-venturi engine
  • Wet/dry capable with a liquid shutoff assembly
  • Backwards compatible with existing pneumatic vacuum components
  • Electrically bonded components and static conductive attachments eliminate static electricity