Learn More
Selleck Chemical LLC B02 10mg
SDP
Supplier:  Selleck Chemical LLC S843410MG
Encompass_Preferred
B02 is a small-molecule inhibitor of human RAD51 with an IC50 of 27.4 ?M, but does not inhibit its E. coli homologue RecA (IC50 > 250 ?M). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3361