Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC B022 | 1202764-53-1 | 99.4% | 100 MG
SDP

Catalog No. 5000234356
Encompass_Preferred

B022 is a selective NF-κB-inducing kinase (NIK) inhibitor research compound commonly used in biochemical and cellular studies targeting NF-κB signaling pathways. It is provided as a high-purity small-molecule suitable for in vitro experiments.

  • Selective inhibition of NF-κB-inducing kinase with low nanomolar potency (Ki ≈ 4.2 nM, IC50 ≈ 15.1 nM).
  • High purity: 99.37% as listed by supplier.
  • Molecular formula C19H16ClN5OS; molecular weight 397.88 g·mol⁻¹.
  • Available in small-molecule research sizes, including 100 MG.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Suitable for studies of inflammation, oxidative stress, and liver injury models.

Catalog No. 50-002-34356 Supplier Medchemexpress LLC Supplier No. HY120501100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

B022 is a selective NF-κB-inducing kinase (NIK) inhibitor research compound commonly used in biochemical and cellular studies targeting NF-κB signaling pathways. It is provided as a high-purity small-molecule suitable for in vitro experiments.

  • Selective inhibition of NF-κB-inducing kinase with low nanomolar potency (Ki ≈ 4.2 nM, IC50 ≈ 15.1 nM).
  • High purity: 99.37% as listed by supplier.
  • Molecular formula C19H16ClN5OS; molecular weight 397.88 g·mol⁻¹.
  • Available in small-molecule research sizes, including 100 MG.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Suitable for studies of inflammation, oxidative stress, and liver injury models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.