Learn More
Biolegend B0306 anti-mouse Hashtag 6 Antibody, 10 ug
Supplier:  Biolegend 155841
Encompass_Preferred
TotalSeq anti-mouse Hashtag reagent is a mixture of two monoclonal antibodies conjugated to the same oligonucleotide. The antibodies are specific against mouse CD45 and MHC class I (of a, b, d, j, k, s, and u haplotypes) and can be used to label hematopoietic and non-hematopoietic cells in most commonly used mouse strains for multiplex single cell sequencing analysis. The conjugated antibodies are pre-mixed to be used following an optimized protocol similar to the CITE-seq workflow.
Catalog No. NC1924575