Learn More
Enzo Life Sciences beta1 integrin (human) monoclonal antibody (DF7) (100µl)
SDP
Supplier:  Enzo Life Sciences BMLIG60610100
Encompass_Preferred
Clone: DF7. Isotype: IgG1. Immunogen: Wheat germ agglutinin binding glycoproteins isolated from human fibroblasts. Species reactivity: Human. Applications: ICC, WB. Purity: Purified.
Catalog No. 50-201-1040