Learn More
MOLECULAR DEPOT LLC B12 Magnetic Beads - 1 mL
Catalog No. 5000013407
Encompass_Preferred
Vitamin B12 Magnetic Beads are a suspension of magnetic particles (about 3 um in diameter 0 1perc solid) conjugated to Vitamin B12 Highly specific and highly stable these particles can be used for the easy one-step capture of various antigens proteins and antibodies specific to Vitamin B12 These particles can be recovered easily using a magnet allowing for various applications such as purification depletion and assay development For in vitro research use only
Catalog No. 50-000-13407 Supplier MOLECULAR DEPOT LLC Supplier No. B2010390
Vitamin B12 Magnetic Beads are a suspension of magnetic particles (about 3 um in diameter 0 1perc solid) conjugated to Vitamin B12 Highly specific and highly stable these particles can be used for the easy one-step capture of various antigens proteins and antibodies specific to Vitamin B12 These particles can be recovered easily using a magnet allowing for various applications such as purification depletion and assay development For in vitro research use only