Learn More
Alkali Scientific Alkali Scientific™, B1201, Micro Bead Sterilizer, with Glass Beads, 115V
SDP
Catalog No. 503029682
Encompass_Preferred
The Alkali Scientific™ Micro Bead Sterilizer B1201 is ideal for the sterilization of small research tools (forceps, scissors, etc.). When the chamber is filled with the included glass beads, high temperatures (up to 300C) can be used to eliminate the presence of bacteria, spores and other microorganisms. Chamber temperature can be viewed on the large LED control panel and conveniently adjusted with the control knob. The outer casing of the unit stays cool to the touch, even when the sterilizer is used all day. The supplied safety cover can be placed over the opening to conserve energy and reduce the heating/ramping rate. The Micro Bead Sterilizer does not use gas, flame or chemicals and is safe for use in hoods. With adjustable temperature, the application range of the sterilizer can also be exanded into incubations and the decontamination of plastic objects/tools. This product is designed for RESEARCH USE ONLY. Not intended for clinical use.
Catalog No. 50-302-9682 Supplier Alkali Scientific Supplier No. RS4201
The Alkali Scientific™ Micro Bead Sterilizer B1201 is ideal for the sterilization of small research tools (forceps, scissors, etc.). When the chamber is filled with the included glass beads, high temperatures (up to 300C) can be used to eliminate the presence of bacteria, spores and other microorganisms. Chamber temperature can be viewed on the large LED control panel and conveniently adjusted with the control knob. The outer casing of the unit stays cool to the touch, even when the sterilizer is used all day. The supplied safety cover can be placed over the opening to conserve energy and reduce the heating/ramping rate. The Micro Bead Sterilizer does not use gas, flame or chemicals and is safe for use in hoods. With adjustable temperature, the application range of the sterilizer can also be exanded into incubations and the decontamination of plastic objects/tools. This product is designed for RESEARCH USE ONLY. Not intended for clinical use.