Learn More
Alkali Scientific Alkali Scientific™ B1202 Tall Micro Bead Sterilizer w/ Glass Beads
SDP
Catalog No. 503029582
Encompass_Preferred
Alkali Scientific™'s B1202 Micro Bead Sterilizer, Glass Bead Sterilizer is ideal for the sterilization of small research tools (forceps, scissors,etc.). When the chamber is filled with the included glass beads, high temperatures (up to 300°C) can be used to eliminate the presence of bacteria, spores and other microorganisms. Supplied with beads. Replacement beads sold separately. For Research Use Only. Not intended for medical or hospital use.
Catalog No. 50-302-9582 Supplier Alkali Scientific Supplier No. RS4202
Alkali Scientific™'s B1202 Micro Bead Sterilizer, Glass Bead Sterilizer is ideal for the sterilization of small research tools (forceps, scissors,etc.). When the chamber is filled with the included glass beads, high temperatures (up to 300°C) can be used to eliminate the presence of bacteria, spores and other microorganisms. Supplied with beads. Replacement beads sold separately. For Research Use Only. Not intended for medical or hospital use.