Learn More
ACROBiosystems Recombinant Protein;Vaccinia Virus B18R Protein;HEK293;500UG;B1R,B18R,VACWR200,Vaccinia Virus,IFNa,IFNb,OCT4,SOX2,KLF4,MYC,OSKM,IFN-alpha,IFN-omega
Supplier:  ACROBiosystems B1RH52H6500UG
Encompass_Preferred
Recombinant Protein;Vaccinia Virus B18R Protein;500UG;Vaccinia Virus B18R Protein, His 20 - Glu 351 (Accession # P25213), was produced in human 293 cells (HEK293) at ACROBiosystems.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized human IFN-α1a at 2μg/mL (100 µL/well),can bind Vaccinia Virus B18R, His Tag (Cat. No. B1R-H52H6) with a linear range of 0.8-10 ng/mL (QC tested).
Catalog No. 50-165-8559