Learn More
Enzo Life Sciences beta2-Microglobulin (human) monoclonal antibody (B2M-01) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804482C100
Encompass_Preferred
Clone: B2M-01. Isotype: IgG2a. Immunogen: Purified human β2-microglobulin. Source: Purified from ascites by DEAE-chromatography and precipitation methods. Species reactivity: Human. Specificity: Recognizes human β2-microglobulin associated with cell-surface MHC Class I molecules and other membrane antigens as well as soluble human β2-microglobulin. Applications: ELISA, Flow Cytometry, IP, WB. Formulation: Liquid. In PBS, pH 7.4, containing 15mM sodium azide.
Catalog No. 50-200-9921