Learn More
Enzo Life Sciences beta2-Microglobulin (human), (native) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX5224610100
Encompass_Preferred
Purified native human β2-Microglobulin protein. Purified native human β2-Microglobulin protein. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In PBS, pH 7.2, containing 150mM sodium chloride, 1mM EDTA sodium salt and 0.05% sodium azide. Source: Purified from urine of patients with chronic renal tubular proteinuria. Long Term Storage: +4°C. Applications: WB .
Catalog No. 50-200-9454