Learn More
Brookfield Engineer Labs General Purpose Oil, B29, Nominal Viscosity 29, 25°C
Catalog No. NC0045321
Encompass
Oil Viscosity Standards are used during calibration checks of specific instruments such as cone/plate viscometers or rheometers as well as Krebs spindle geometry instruments.General Purpose Oil Fluids are best suited for those instruments using Cone/Plate Viscometers/Rheometers.Accuracy: +/-1% of viscosity valueAppropriate for use at shear rates greater than 500 sec-1Recommended for use with cone/plate Viscometers at viscosities above 5,000 cPBrookfield oil viscosity standards are hydrocarbon based, either mineral oil or polybutenes Note: Other oil fluids are available - call for detailsBrookfield Viscosity Standards are accurate to ±1% of the stated viscosity and are certified by methods traceable to the United States National Institute of Standards and Technology (NIST).
Catalog No. NC0045321 Supplier Brookfield Engineer Labs Supplier No. B29
Oil Viscosity Standards are used during calibration checks of specific instruments such as cone/plate viscometers or rheometers as well as Krebs spindle geometry instruments.General Purpose Oil Fluids are best suited for those instruments using Cone/Plate Viscometers/Rheometers.Accuracy: +/-1% of viscosity valueAppropriate for use at shear rates greater than 500 sec-1Recommended for use with cone/plate Viscometers at viscosities above 5,000 cPBrookfield oil viscosity standards are hydrocarbon based, either mineral oil or polybutenes Note: Other oil fluids are available - call for detailsBrookfield Viscosity Standards are accurate to ±1% of the stated viscosity and are certified by methods traceable to the United States National Institute of Standards and Technology (NIST).