Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 567MSM2XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM617. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HL-60 or HeLa cells. Melanomas, Lymphoma. Carcinoma of Stomach, Cervix, Endometrial, Kidney or Colon. Cellular Staining: Cell Surface, Extracellular (Secreted). Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Full length recombinant human B2M protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 12kDa.Chromosome: 15q21-q22.2. Gene Symbol: B2M. Gene ID: 567. Ensemble: ENSG00000166710. Uniprot: XP_005254606.
Catalog No. 50-188-5086