Learn More
Sino Biological B2M / beta-2 microglobulin Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11976MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human B2M / beta-2 microglobulin (rh B2M / beta-2 microglobulin; Catalog#11976-H08H; NP_004039.1; Met1-Met119). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8488