Learn More
Brady™ B30 Printer Series Replacement Media Wiper Pad
Catalog No. 17607624 Shop All Brady Corporation Products
Catalog No. 17-607-624 Supplier Brady™ Supplier No. 142114
  • $51.60
    $47.65 / Pack of 5
    Save $3.95
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Keep your Brady printer in top working condition with these replacement media wiper pads.
Keep your Brady printer in top working condition with these replacement media wiper pads. Package includes five media wiper pads (printer sold separately).
  • For use with BBP™30, S3100, BBP™31, and BBP™33 BradyPrinters.
  • Essential for keeping continuous label media clean and free of debris that can cause print voids.
  • Cleans the label tape surface before it passes through the thermal transfer printhead.
Specifications
Product Type Cleaning Kit - Wiper
Autoclavable Not Autoclavable
Sterility Non-sterile