Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ B30 Series Raised Profile Rectangular Labels

Catalog No. 17111239 Shop All Brady Corporation Products
Catalog No. 17-111-239 Supplier Brady™ Supplier No. B30EP174593WT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Raised panel adhesive taped polyester (B-593) labels are designed for patch panel identification, external push-buttons, switches and internal connection points.

  • Raised panel Adhesive-Taped Polyester (B-593) labels are designed for patch panel identification, external push-buttons, switches and internal connection points.
  • 1 in H x 4 in W White 300/RL
  • Can be used as rating and serial plates using alphanumerics that require nameplate quality.
  • Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags.
  • UL approved. Service temperature of -4°F to 212°F (-20°C to 100°C). A ribbon is required to print this material. Use Part Number B30-R6000.

Specifications

Adhesive Type Permanent Acrylic
Coating Polyester
Color White
Finish Gloss
Format Cartridge (Label)
Material Polyester
Print Technology Thermal Transfer
Sterility Non-sterile
For Use With (Application) Electrical Labeling, General Identification
Length (English) 1 in.
Width (English) 4.25 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.