Learn More
ATCC B35; Neuroblastoma; Rat
Catalog No. 502382908
Encompass
B35 is a neuronal neuroblast cell line that was isolated from the central nervous system of a 4-to-10-month-old, rat with neuroblastoma. This cell line was deposited by P Maness and can be used in neuroscience research.
Catalog No. 50-238-2908 Supplier ATCC Supplier No. CRL2754
B35 is a neuronal neuroblast cell line that was isolated from the central nervous system of a 4-to-10-month-old, rat with neuroblastoma. This cell line was deposited by P Maness and can be used in neuroscience research.