Learn More
Alkali Scientific Alkali Scientific™ B3D1020 Mini Mixer (3D) mixer, Fixed Speed, Small Platform
SDP
Catalog No. 503029646
Encompass_Preferred
The Alkali Scientific™ B3D1020 Mini-Mixer is multi-functional shaker provides a 3-dimensional "nutating" motion for thorough, foam-free mixing in blood and centrifuge tubes, as well as in a variety of flat vessels. The Alkali Scientific™ B3D1020 Mini-Mixer comes standard with autoclavable dimpled mats while optional flat mats are also available for mixing other vessel types. Stacking platforms can be purchased as accessories, effectively doubling the usable surface areal. Includes dimpled mat; ideal for mixing in blood & centrifuge tubes Tri-directional motion for thorough, uniform mixing Optional stacking platforms available .
Catalog No. 50-302-9646 Supplier Alkali Scientific Supplier No. RS7011
The Alkali Scientific™ B3D1020 Mini-Mixer is multi-functional shaker provides a 3-dimensional "nutating" motion for thorough, foam-free mixing in blood and centrifuge tubes, as well as in a variety of flat vessels. The Alkali Scientific™ B3D1020 Mini-Mixer comes standard with autoclavable dimpled mats while optional flat mats are also available for mixing other vessel types. Stacking platforms can be purchased as accessories, effectively doubling the usable surface areal. Includes dimpled mat; ideal for mixing in blood & centrifuge tubes Tri-directional motion for thorough, uniform mixing Optional stacking platforms available .