Learn More
Alkali Scientific Alkali Scientific™ B3D1308 BlotBoy™ 3D Rocker w/ Flat Mat, 115V, 50/60 Hz, 12 RPM
SDP
Catalog No. 503029647
Encompass_Preferred
Modeled after our popular Alkali Scientific™ BioMixer™ series of "nutating" shakers, the Alkali Scientific™ B3D1308 BlotBoy™ is a platform rocker that provides the perfect 3-dimensional motion for all blot related applications.The tilt angle and speed have been optimally set for gentle, but thorough mixing in gel trays, boxes and other flat vessels. The Alkali Scientific™ B3D1308 BlotBoy™ orbital shaker is supplied with an autoclavable flat mat and is ideal for blot washes and gel staining/destaining. The Alkali Scientific™ B3D1308 BlotBoy™ is supplied with a 12 x 12 inch platform. The Alkali Scientific™ B3D1308 BlotBoy™ can accept stackable platforms and handle loads up to 2kg. Comes with maintenance free brushless motor and is safe for use in cold rooms and incubators.
Catalog No. 50-302-9647 Supplier Alkali Scientific Supplier No. RS7035
Modeled after our popular Alkali Scientific™ BioMixer™ series of "nutating" shakers, the Alkali Scientific™ B3D1308 BlotBoy™ is a platform rocker that provides the perfect 3-dimensional motion for all blot related applications.The tilt angle and speed have been optimally set for gentle, but thorough mixing in gel trays, boxes and other flat vessels. The Alkali Scientific™ B3D1308 BlotBoy™ orbital shaker is supplied with an autoclavable flat mat and is ideal for blot washes and gel staining/destaining. The Alkali Scientific™ B3D1308 BlotBoy™ is supplied with a 12 x 12 inch platform. The Alkali Scientific™ B3D1308 BlotBoy™ can accept stackable platforms and handle loads up to 2kg. Comes with maintenance free brushless motor and is safe for use in cold rooms and incubators.