Learn More
Aviva Systems Biology Corp B3galnt1 Antibody - C-terminal region (ARP44857_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44857_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B3galnt1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: B3galt3, B3galnt1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-164