Learn More
Aviva Systems Biology Corp B3GALNT1 Antibody - middle region (ARP44858_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44858_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B3GALNT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human B3GALNT1. Alternate names: B3GALT3, GLCT3, GLOB, Gb4Cer, P, P1, beta3Gal-T3, galT3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58918