Learn More
Aviva Systems Biology Corp B3GALNT2 Antibody - middle region (ARP49061_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49061_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B3GALNT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human B3GALNT2. Alternate names: B3GalNAc-T2, MGC39558ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60006