Learn More
Aviva Systems Biology Corp B3GALT1 Antibody - N-terminal region (ARP49606_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49606_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B3GALT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human B3GALT1. Alternate names: MGC126594, beta3Gal-T1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60122